Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine
4014948.0005
4014948.0025
4014948.0100
B-1785.0005BAEA
180.07
CAD
B-1785.0005BA
B-1785.0025BA
B-1785.0100BA
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
Sequence: Fmoc-Asn(Trt)-OH
For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".
Formula:
C₃₈H₃₂N₂O₅ MW: 596.68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077049 CAS Number: 132388-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...