Fmoc-Abu-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-(Fmoc-amino)butyric acid

4006449.0001 4006449.0005 4006449.0025
B-1780.0001BAEA 180.91 CAD
B-1780.0001BA B-1780.0005BA B-1780.0025BA
Fmoc-Abu-OH
Fmoc-Abu-OH

Sequence: Fmoc-Abu-OH

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080268
CAS Number: 135112-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR