Fmoc-O-ethyl-D-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Tyr(4-Et)-OH, Fmoc-D-Tyr(Et)-OH

4015247.0001 4015247.0005
B-1775.0001BAEA 439.41 CAD
B-1775.0001BA B-1775.0005BA
Fmoc-O-ethyl-D-tyrosine
Fmoc-O-ethyl-D-tyrosine

Sequence: Fmoc-D-Tyr(Et)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
p-Methoxyphenylalanine and p-ethoxyphenylalanine.


Synonym(s): Fmoc-4-ethoxy-D-Phe-OH

Formula: C₂₆H₂₅NO₅
Storage Temperature: Ambient
MDL Number: MFCD00237035
CAS Number: 162502-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR