Fmoc-β-Ala-OPFP

Supplier: BACHEM AMERICAS INC

4012057.0005 4012057.0025
B-1765.0005BAEA 439.41 CAD
B-1765.0005BA B-1765.0025BA
Fmoc-β-Ala-OPFP
Formula: C₂₄H₁₆F₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD00237655
CAS Number: 149303-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR