Fmoc-Thr-OH

Supplier: BACHEM AMERICAS INC

4015218.0005 4015218.0025
B-1745.0005BAEA 180.14 CAD
B-1745.0005BA B-1745.0025BA
Fmoc-Thr-OH
Fmoc-Thr-OH

Sequence: Fmoc-Thr-OH


Bachem also offers phosphothreonine derivatives.


Educt for the synthesis of O-glycosylated threonine building blocks. Liu et al. described the synthesis of O-mannosylated Fmoc-Thr.

Formula: C₁₉H₁₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD00077072
CAS Number: 73731-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR