Fmoc-3-iodo-L-Tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-3-iodo-Tyr-OH, Fmoc-Tyr(3-I)-OH

4015214.0001 4015214.0005
B-1740.0001BAEA 828.77 CAD
B-1740.0001BA B-1740.0005BA
Fmoc-3-iodo-L-Tyrosine
Fmoc-3-iodo-L-Tyrosine

Sequence: Fmoc-3-iodo-Tyr-OH


Bachem offers derivatives of (di)halogenated tyrosines, (di)nitrotyrosine and other 3-substituted tyrosines.
See also 3-Methoxytyramine · HCl (Q-1330).

Formula: C₂₄H₂₀INO₅
MW: 529.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00235906
CAS Number: 134486-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR