Fmoc-Sar-OH
Supplier: BACHEM AMERICAS INC
4009790.0005
4009790.0025
4009790.0100
B-1720.0005BAEA
185.46
CAD
B-1720.0005BA
B-1720.0025BA
B-1720.0100BA
Fmoc-Sar-OH
Fmoc-Sar-OH
Sequence: Fmoc-Sar-OH
Bachem also offers Fmoc-Sar-Wang resin (D-1805) and Fmoc-Sar-SASRIN resin (-D-1645).
Synonym(s): Fmoc-N-Me-Gly-OH
Formula:
C₁₈H₁₇NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00151926 CAS Number: 77128-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...