N-Fmoc-N'-trityl-D-histidine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-His(1-Trt)-OH, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-D-histidine, Fmoc-D-His(Trt)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-Fmoc-1-trityl-D-histidine, Nalpha-Fmoc-tele-trityl-D-histidine

4014118.0001 4014118.0005 4014118.0025
B-1710.0001BAEA 175.66 CAD
B-1710.0001BA B-1710.0005BA B-1710.0025BA
N-Fmoc-N'-trityl-D-histidine
N-Fmoc-N'-trityl-D-histidine

Sequence: Fmoc-D-His(1-Trt)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-D-His(τ-Trt)-OH

Formula: C₄₀H₃₃N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD00077061
CAS Number: 135610-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR