Fmoc-D-Arg(Pmc)-OH

Supplier: BACHEM AMERICAS INC

4014141.0001 4014141.0005 4014141.0025
B-1680.0001BAEA 250.43 CAD
B-1680.0001BA B-1680.0005BA B-1680.0025BA
Fmoc-D-Arg(Pmc)-OH
Fmoc-D-Arg(Pmc)-OH

Sequence: Fmoc-D-Arg(Pmc)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₅H₄₂N₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD00065619
CAS Number: 157774-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR