Fmoc-Arg(Pmc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: FMOC-L-arginine(PMC)

4013349.0001 4013349.0005 4013349.0025 4013349.0100
B-1670.0001BAEA 175.66 CAD
B-1670.0001BA B-1670.0005BA B-1670.0025BA B-1670.0100BA
Fmoc-Arg(Pmc)-OH
Fmoc-Arg(Pmc)-OH

Sequence: Fmoc-Arg(Pmc)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₅H₄₂N₄O₇S
MW: 662.802 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065618
CAS Number: 119831-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR