Fmoc-Thi-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-3-(2-thienyl)-L-alanine, Fmoc-β-(2-thienyl)-Ala-OH, (S)-2-(Fmoc-amino)-3-(2-thienyl)propionic acid, N-α-Fmoc-β-thienyl-L-alanine

4009752.0001 4009752.0005
B-1665.0001BAEA 542.13 CAD
B-1665.0001BA B-1665.0005BA
Fmoc-Thi-OH
Fmoc-Thi-OH

Sequence: Fmoc-β-(2-thienyl)-Ala-OH
Synonym(s): Fmoc-Thi-OH


2-Thi derivatives.


Fmoc-Thi-OH.

Formula: C₂₂H₁₉NO₄S
MW: 393.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065676
CAS Number: 130309-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR