Fmoc-3,4-dehydroPro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2,5-dihydro-1H-pyrrole-2-carboxylic acid, Fmoc-3,4-dehydro-Pro-OH

4013705.0001 4013705.0005
B-1660.0001BAEA 2039.53 CAD
B-1660.0001BA B-1660.0005BA
Fmoc-3,4-dehydroPro-OH
Fmoc-3,4-dehydroPro-OH

Sequence: Fmoc-3,4-dehydro-Pro-OH


Dhp derivatives.

Formula: C₂₀H₁₇NO₄
MW: 335.359 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151940
CAS Number: 135837-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR