(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoate

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-His(1-Trt)-OPfp, Fmoc-His(Trt)-OPfp

4011864.0001 4011864.0005 4011864.0025
B-1650.0001BAEA 201.47 CAD
B-1650.0001BA B-1650.0005BA B-1650.0025BA
(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoate
(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoate

Sequence: Fmoc-His(1-Trt)-OPfp


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-His(τ-Trt)-OPfp

Formula: C₄₆H₃₂F₅N₃O₄
MW: 785.771 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065653
CAS Number: 109434-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR