Fmoc-D-CYS(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Cys(tBu)-OH

4011898.0001 4011898.0005
B-1645.0001BAEA 1406.71 CAD
B-1645.0001BA B-1645.0005BA
Fmoc-D-CYS(tBu)-OH
Fmoc-D-CYS(tBu)-OH

Sequence: Fmoc-D-Cys(tBu)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).

Formula: C₂₂H₂₅NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00077052
CAS Number: 131766-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR