Fmoc-D-isoleucine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-D-Ile-OH, (2R,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylpentanoic acid
4011893.0001
4011893.0005
B-1630.0001BAEA
770.52
CAD
B-1630.0001BA
B-1630.0005BA
Fmoc-D-isoleucine
Fmoc-D-isoleucine
Sequence: Fmoc-D-Ile-OH
Formula:
C₂₁H₂₃NO₄ MW: 353.418 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00080291 CAS Number: 143688-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...