Fmoc-D-Pro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-D-proline

4011894.0005 4011894.0025
B-1625.0005BAEA 351.47 CAD
B-1625.0005BA B-1625.0025BA
Fmoc-D-Pro-OH
Fmoc-D-Pro-OH

Sequence: Fmoc-D-Pro-OH

Formula: C₂₀H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00077067
CAS Number: 101555-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR