(2R,2'R)-3,3'-Disulfanediylbis(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid)

Supplier: BACHEM AMERICAS INC

Synonyms: (Fmoc-Cys-OH)₂

4011808.0001 4011808.0005 4011808.0025
B-1615.0001BAEA 180.91 CAD
B-1615.0001BA B-1615.0005BA B-1615.0025BA
(2R,2'R)-3,3'-Disulfanediylbis(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid)
(2R,2'R)-3,3'-Disulfanediylbis(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid)

Sequence: (Fmoc-Cys-OH)₂


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.

Formula: C₃₆H₃₂N₂O₈S₂
MW: 684.778 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237447
CAS Number: 135273-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR