Fmoc-Lys(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

4011122.0005 4011122.0025 4011122.0100
B-1610.0005BAEA 206.51 CAD
B-1610.0005BA B-1610.0025BA B-1610.0100BA
Fmoc-Lys(Fmoc)-OH
Fmoc-Lys(Fmoc)-OH

Sequence: Fmoc-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Building block for the synthesis of branched peptides and MAP matrices.

Formula: C₃₆H₃₄N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00080275
CAS Number: 78081-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR