Fmoc-Tyr(SO3H)-OH

Supplier: BACHEM AMERICAS INC

4029106.0001 4029106.0005
B-1600.0001BAEA 916.53 CAD
B-1600.0001BA B-2765.0001BA B-2765.0005BA
Fmoc-Tyr(SO3H)-OH
Fmoc-Tyr(SO3H)-OH

Sequence: Fmoc-Tyr(SO₃H)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Bachem also offers a protected stable sulfotyrosine mimic, Fmoc-4-sulfomethyl-Phe(Tce)-OH (B-4110).


Synonym(s): Fmoc-sulfotyrosine sodium salt

Formula: C₂₄H₂₁NO₈S
Storage Temperature: Freezer
MDL Number: MFCD00077078
CAS Number: 181952-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR