Fmoc-Glu-OtBu
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid, N-Fmoc-L-glutamic acid 1-tert-butyl ester, Fmoc-Glu-OtBu
4011877.0001
4011877.0005
4011877.0025
B-1595.0001BAEA
232.35
CAD
B-1595.0001BA
B-1595.0005BA
B-1595.0025BA
Fmoc-Glu-OtBu
Fmoc-Glu-OtBu
Sequence: Fmoc-Glu-OtBu
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₂₄H₂₇NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00065648 CAS Number: 84793-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...