Fmoc-D-glutamine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-D-Gln-OH, N-Fmoc-D-glutamine
4011842.0001
4011842.0005
4011842.0025
B-1585.0001BAEA
175.66
CAD
B-1585.0001BA
B-1585.0005BA
B-1585.0025BA
Fmoc-D-glutamine
Fmoc-D-glutamine
Sequence: Fmoc-D-Gln-OH
For isoglutamine derivatives please see the family "Glutamic Acid".
Formula:
C₂₀H₂₀N₂O₅ MW: 368.389 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00062959 CAS Number: 112898-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...