N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Tyr-OH, N-Fmoc-L-tyrosine

4012011.0001 4012011.0005
B-1580.0001BAEA 358.5 CAD
B-1580.0001BA B-1580.0005BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine

Sequence: Fmoc-Tyr-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₄H₂₁NO₅
MW: 403.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00134890
CAS Number: 92954-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR