N-ɑ-Fmoc-N-Trityl-L-histidine

Supplier: BACHEM AMERICAS INC

Synonyms: N-ɑ-Fmoc-tele-trityl-L-histidine, Fmoc-His(Trt)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-ɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-L-histidine, N-Fmoc-1-trityl-L-histidine

4011444.0005 4011444.0025 4011444.0100
B-1570.0005BAEA 175.66 CAD
B-1570.0005BA B-1570.0025BA B-1570.0100BA
N-ɑ-Fmoc-N-Trityl-L-histidine
N-alpha-Fmoc-N-Trityl-L-histidine

Sequence: Fmoc-His(1-Trt)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-His(τ-Trt)-OH

Formula: C₄₀H₃₃N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD00043332
CAS Number: 109425-51-6
EINECS: 439-640-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR