Fmoc-6-Ahx-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-ε-aminocaproic acid, Fmoc-ε-Acp-OH, 6-(Fmoc-amino)hexanoic acid

4006429.0001 4006429.0005 4006429.0025
B-1560.0001BAEA 180.91 CAD
B-1560.0001BA B-1560.0005BA B-1560.0025BA
Fmoc-6-Ahx-OH
Fmoc-6-Ahx-OH

Sequence: Fmoc-ε-aminocaproic acid


Flexible spacer molecules of various length.
A flexible non-polar spacer moiety abbreviated as LC, εAca, or εAhx. Bachem also offers Fmoc-εAhx-Wang resin (D-1050).


Synonym(s): Fmoc-LC-OH#Fmoc-εAhx-OH#Fmoc-6-aminohexanoic acid

Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00077045
CAS Number: 88574-06-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR