Fmoc-L-homophenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-HoPhe-OH, (S)-2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-phenylbutyric acid, Fmoc-Homophe-OH, N-Fmoc-L-homophenylalanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine, (S)-2-(Fmoc-amino)-4-phenylbutyric acid

4011602.0001 4011602.0005
B-1535.0001BAEA 309.81 CAD
B-1535.0001BA B-1535.0005BA
Fmoc-L-homophenylalanine
Fmoc-L-homophenylalanine

Sequence: Fmoc-Homophe-OH


Hph derivatives. Bachem also offers Fmoc-Hph-Wang resin, D-2005.


Synonym(s): (S)-2-(Fmoc-amino)-4-phenyl-butyric acid#NPC-14686

Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077046
CAS Number: 132684-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR