Fmoc-Cys(StBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-S-(tert-butylthio)-L-cysteine

4007445.0005 4007445.0025 4007445.0100
B-1530.0005BAEA 418.62 CAD
B-1530.0005BA B-1530.0025BA B-1530.0100BA
Fmoc-Cys(StBu)-OH
Fmoc-Cys(StBu)-OH

Sequence: Fmoc-Cys(StBu)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).


Was converted into the corresponding N-Me-Cys derivative in excellent yield. Educt for obtaining the Cys 4-methyl-2,6,7-trioxabicyclo[2,2,2]octyl (OBO) orthoester. The OBO ester was attached to a thiol resin as disulfide for obtaining peptides containing a C-terminal cysteine.

Formula: C₂₂H₂₅NO₄S₂
MW: 431.565 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042617
CAS Number: 73724-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR