trans-N-Fmoc-4-tert-butoxy-L-proline

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-4-trans-(tert-butoxy)-L-proline, Fmoc-Hyp(tBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline, (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxy)pyrrolidine-2-carboxylic acid

4008172.0001 4008172.0005 4008172.0025
B-1525.0001BAEA 180.91 CAD
B-1525.0001BA B-1525.0005BA B-1525.0025BA
trans-N-Fmoc-4-tert-butoxy-L-proline
trans-N-Fmoc-4-tert-butoxy-L-proline

Sequence: Fmoc-Hyp(tBu)-OH


Hyp denotes L-trans-hydroxyproline.

Formula: C₂₄H₂₇NO₅
MW: 409.482 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151930
CAS Number: 122996-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR