Fmoc-Gly-Gly-Gly-OH
Supplier: BACHEM AMERICAS INC
4003169.0001 4003169.0005
B-1490.0001BAEA 407.01 CAD
B-1490.0001BA B-1490.0005BA
Fmoc-Gly-Gly-Gly-OH
Proteins and Peptides
For Fmoc-dipeptides Fmoc-Xaa-Gly-OH and Fmoc-Xaa-Pro-OH, which can be used as building blocks in Fmoc-SPPS, please see also the subfamily 'Fmoc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. Pseudoproline dipeptides and isoacyl dipeptides can be found in this section as well. In case of a strong tendency towards diketopiperazine formation during Fmoc-SPPS, Fmoc-dipeptides have been coupled even at the risk of a some epimerization.
Fmoc-triglycine.