Fmoc-val-OSu

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-valine N-succinimidyl ester, Fmoc-Val-OSu

4003501.0005 4003501.0025
B-1465.0005BAEA 264.68 CAD
B-1465.0005BA B-1465.0025BA
Fmoc-val-OSu
Fmoc-val-OSu

Sequence: Fmoc-Val-OSu


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₂₄H₂₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00153370
CAS Number: 130878-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR