N-Fmoc-D-valine

Supplier: BACHEM AMERICAS INC

4007570.0005 4007570.0025
B-1460.0005BAEA 299.43 CAD
B-1460.0005BA B-1460.0025BA
N-Fmoc-D-valine
N-Fmoc-D-valine

Sequence: Fmoc-D-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₂₀H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00062953
CAS Number: 84624-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR