N-Fmoc-L-valine

Supplier: BACHEM AMERICAS INC

4093145.0025 4093145.0100 4093145.0500
B-1455.0025BAEA 184.64 CAD
B-1455.0025BA B-1455.0100BA B-1455.0500BA
N-Fmoc-L-valine
N-Fmoc-L-valine

Sequence: Fmoc-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₂₀H₂₁NO₄
MW: 339.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037124
CAS Number: 68858-20-8
EINECS: 272-515-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR