N(α)-Fmoc-L-tryptophan
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Trp-OH
4003185.0025
4003185.0100
4003185.0500
B-1445.0025BAEA
175.66
CAD
B-1445.0025BA
B-1445.0100BA
B-1445.0500BA
N(α)-Fmoc-L-tryptophan
N(α)-Fmoc-L-tryptophan
Sequence: Fmoc-Trp-OH
Formula:
C₂₆H₂₂N₂O₄ MW: 426.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037126 CAS Number: 35737-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...