N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Cys(Trt)-OH, N-Fmoc-S-trityl-L-cysteine
4007272.0005
4007272.0025
4007272.0100
B-1440.0005BAEA
180.07
CAD
B-1440.0005BA
B-1440.0025BA
B-1440.0100BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
Sequence: Fmoc-Cys(Trt)-OH
Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).
Standard derivative for Fmoc-SPPS. Educt for obtaining S-alkylated cysteines via thiol-ene reaction.
Formula:
C₃₇H₃₁NO₄S MW: 585.72 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038538 CAS Number: 103213-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...