Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine

Supplier: BACHEM AMERICAS INC

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-tosyl-L-arginine, Nalpha-Fmoc-Nomega-tosyl-L-arginine, Nα-Fmoc-Nω-tosyl-L-arginine, Fmoc-Arg(Tos)-OH

4004138.0005 4004138.0025
B-1435.0005BAEA 206.44 CAD
B-1435.0005BA B-1435.0025BA
Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine
Nα-Fmoc-Nω-(p-toluenesulfonyl)-L-arginine

Sequence: Fmoc-Arg(Tos)-OH


Fmoc-Arg(Tos) was shown to inhibit HIV-1 protease (IC₅₀ 5 μM) and cathepsin D (IC₅₀ 12 μM).

Formula: C₂₈H₃₀N₄O₆S
MW: 550.63 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063331
CAS Number: 83792-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR