Fmoc-Pro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

4003150.0025 4003150.0100 4003150.0500
B-1420.0025BAEA 182.36 CAD
B-1420.0025BA B-1420.0100BA B-1420.0500BA
Fmoc-Pro-OH
Fmoc-Pro-OH

Sequence: Fmoc-Pro-OH

Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037122
CAS Number: 71989-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR