Fmoc-D-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine

4005952.0005 4005952.0025
B-1410.0005BAEA 227.28 CAD
B-1410.0005BA B-1410.0025BA
Fmoc-D-phenylalanine
Fmoc-D-phenylalanine

Sequence: Fmoc-D-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₂₄H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00062955
CAS Number: 86123-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR