Fmoc-L-phenylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine
4100450.0025
4100450.0100
4100450.0500
B-1405.0025BAEA
175.66
CAD
B-1405.0025BA
B-1405.0100BA
B-1405.0500BA
Fmoc-L-phenylalanine
Fmoc-L-phenylalanine
Sequence: Fmoc-Phe-OH
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Formula:
C₂₄H₂₁NO₄ MW: 387.4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037128 CAS Number: 35661-40-6 EINECS: 252-661-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...