N-Fmoc-L-norleucine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Nle-OH, FMOC-L-Norleucine
4004485.0005
4004485.0025
B-1400.0005BAEA
180.91
CAD
B-1400.0005BA
B-1400.0025BA
N-Fmoc-L-norleucine
N-Fmoc-L-norleucine
Sequence: Fmoc-Nle-OH
Synonym(s): Fmoc-L-2-aminohexanoic acid
Bachem also offers Fmoc-Nle-Wang resin (D-1795) and Fmoc-Nle-SASRIN resin (D-1730).
Formula:
C₂₁H₂₃NO₄ MW: 353.4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037537 CAS Number: 77284-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...