Fmoc-N-Me-Val-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-valine, Fmoc-N-Me-Val-OH, N-Fmoc-N-methyl-L-valine

4009586.0001 4009586.0005
B-1380.0001BAEA 258.09 CAD
B-1380.0001BA B-1380.0005BA
Fmoc-N-Me-Val-OH
Fmoc-N-Me-Val-OH

Sequence: Fmoc-N-Me-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
MeVal, αMeVal.


The acid fluoride of this sterically hindered amino acid could be coupled to N-methylated amino acids in HFIP at 55°C.

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153395
CAS Number: 84000-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR