Fmoc-Cys(pMeOBzl)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, Fmoc-Cys(Mob)-OH

4007329.0005 4007329.0025
B-1375.0005BAEA 276.63 CAD
B-1375.0005BA B-1375.0025BA
Fmoc-Cys(pMeOBzl)-OH
Fmoc-Cys(pMeOBzl)-OH

Sequence: Fmoc-Cys(Mob)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).

Formula: C₂₆H₂₅NO₅S
MW: 463.548 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065635
CAS Number: 141892-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR