N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine

Supplier: BACHEM AMERICAS INC

4004745.0005 4004745.0025
B-1365.0005BAEA 351.38 CAD
B-1365.0005BA B-1365.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine

Sequence: Fmoc-D-Met-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.

Formula: C₂₀H₂₁NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00062958
CAS Number: 112883-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR