N-Fmoc-L-leucine

Supplier: BACHEM AMERICAS INC

4003139.0025 4003139.0100 4003139.0500
B-1345.0025BAEA 184.64 CAD
B-1345.0025BA B-1345.0100BA B-1345.0500BA
N-Fmoc-L-leucine
N-Fmoc-L-leucine

Sequence: Fmoc-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).


Synonym(s): NPC 15199

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037133
CAS Number: 35661-60-0
EINECS: 252-662-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR