N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine

Supplier: BACHEM AMERICAS INC

4003168.0025 4003168.0100 4003168.0500
B-1340.0025BAEA 180.14 CAD
B-1340.0025BA B-1340.0100BA B-1340.0500BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine

Sequence: Fmoc-Ile-OH

Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037125
CAS Number: 71989-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR