N-Fmoc-glycine

Supplier: BACHEM AMERICAS INC

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

4003078.0025 4003078.0100 4003078.0500
B-1330.0025BAEA 180.14 CAD
B-1330.0025BA B-1330.0100BA B-1330.0500BA
N-Fmoc-glycine
N-Fmoc-glycine

Sequence: Fmoc-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₁₇H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037140
CAS Number: 29022-11-5
EINECS: 249-373-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR