N-Fmoc-D-glutamic acid 5-tert-butyl ester

Supplier: BACHEM AMERICAS INC

Synonyms: 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid

4004498.0001 4004498.0005 4004498.0025
B-1320.0001BAEA 175.66 CAD
B-1320.0001BA B-1320.0005BA B-1320.0025BA
N-Fmoc-D-glutamic acid 5-tert-butyl ester
N-Fmoc-D-glutamic acid 5-tert-butyl ester

Sequence: Fmoc-D-Glu(OtBu)-OH · H₂O CAS-No.: 104091-08-9 Mol.weight: 443.50 SumFormula: C₂₄H₂₇NO₆ · H₂O Bulk item no.: B-1320 Synonym(s): EINECS no.: - MDL no.: MFCD00797526


Sequence: Fmoc-D-Glu(OtBu)-OH · H₂O


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₂₄H₂₇NO₆
MW: 425.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077055
CAS Number: 104091-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR