Fmoc-Glu(OtBu)-OH·H₂O

Supplier: BACHEM AMERICAS INC

4003082.0025 4003082.0100 4003082.0500
B-1315.0025BAEA 369.02 CAD
B-1315.0025BA B-1315.0100BA B-1315.0500BA
Fmoc-Glu(OtBu)-OH·H₂O
Fmoc-Glu(OtBu)-OH·H₂O

Sequence: Fmoc-Glu(OtBu)-OH · H₂O


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


CAS-Number (anhydrous) 71989-18-9.

Formula: C₂₄H₂₉NO₇
Storage Temperature: Refrigerator
MDL Number: MFCD00150485
CAS Number: 204251-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR