Fmoc-Glu(OBzl)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Fmoc-L-glutamic acid 5-benzyl ester
4004137.0005
4004137.0025
B-1310.0005BAEA
232.35
CAD
B-1310.0005BA
B-1310.0025BA
Fmoc-Glu(OBzl)-OH
Fmoc-Glu(OBzl)-OH
Sequence: Fmoc-Glu(OBzl)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₂₇H₂₅NO₆ MW: 459.498 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065642 CAS Number: 123639-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...