Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Fmoc-L-glutamine, Nɑ-Fmoc-L-glutamine, Fmoc-Gln-OH

4003156.0025 4003156.0100
B-1300.0025BAEA 175.66 CAD
B-1300.0025BA B-1300.0100BA
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine

Sequence: Fmoc-Gln-OH


For isoglutamine derivatives please see the family "Glutamic Acid".

Formula: C₂₀H₂₀N₂O₅
MW: 368.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037137
CAS Number: 71989-20-3
EINECS: 276-254-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR