Fmoc-3,5-dibromo-L-Tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-3,5-dibromo-Tyr-OH

4008952.0001 4008952.0005
B-1275.0001BAEA 180.91 CAD
B-1275.0001BA B-1275.0005BA
Fmoc-3,5-dibromo-L-Tyrosine
Fmoc-3,5-dibromo-L-Tyrosine

Sequence: Fmoc-3,5-dibromo-Tyr-OH


Bachem offers derivatives of (di)halogenated tyrosines, (di)nitrotyrosine and other 3-substituted tyrosines.
3-Bromotyrosine and 3,5-dibromotyrosine are major products of protein oxidation by eosinophil peroxidase utilizing bromide ions present in plasma.

Formula: C₂₄H₁₉Br₂NO₅
MW: 561.226 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065686
CAS Number: 201484-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR