Fmoc-Lys(Z)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysine, Fmoc-Lys(Cbz)-OH, Fmoc-Lys(Z)-OH, Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, Nalpha-Fmoc-Nepsilon-Cbz-L-lysine

4004133.0005 4004133.0025
B-1270.0005BAEA 180.91 CAD
B-1270.0005BA B-1270.0025BA
Fmoc-Lys(Z)-OH
Fmoc-Lys(Z)-OH

Sequence: Fmoc-Lys(Z)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Building block for Lys-containing short polar peptides. They are easier to purify, because Z-protection renders them more lipophilic.

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065662
CAS Number: 86060-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR